Tetraethylammonium perchlorate ≥98% (dry weight) wetted with 10% water

Supplier: Thermo Scientific Chemicals
Danger

44062-09 44062-18
CAAA44062-09EA 103.81 CAD
CAAA44062-09 CAAA44062-18
Tetraethylammonium perchlorate ≥98% (dry weight) wetted with 10% water
Tetraethylammonium perchlorate

Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.


Purity: 98% CAS No.: 2567-83-1 Molecular Formula: (C2H5)4NClO4 Formula Weight: 229.71 Physical Form: Wet crystals Odor: Odorless Beilstein Reference: 3638093 UN No.: 1479 MDL No.: MFCD00041977

Formula: C₈H₂₀ClNO₄
Melting Pt: >300 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041977
CAS Number: 2567-83-1
EINECS: 219-904-3
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR