Sodium tetrachloropalladate(II) hydrate (≥30% Pd) ≥99.999% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate

43945-03 43945-06
CAAA43945-03EA 265.44 CAD
CAAA43945-03 CAAA43945-06
Sodium tetrachloropalladate(II) hydrate (≥30% Pd) ≥99.999% (metals basis), Premion®
Sodium tetrachloropalladate(II) hydrate

Purity: 99.999% CAS No.: 13820-53-6 Molecular Formula: Cl4Na2Pd.xH2O (x≈3) Formula Weight: 294.20(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red-brown MDL No.: MFCD00003487

Formula: Cl₄Na₂Pd·xH₂O
MW: 294.21 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00003487
CAS Number: 13820-53-6
EINECS: 237-502-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR