Potassium heptafluorotantalate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

43733-09 43733-18 43733-30
CAAA43733-09EA 70.31 CAD
CAAA43733-09 CAAA43733-18 CAAA43733-30
Potassium heptafluorotantalate ≥99.99% (metals basis)
Potassium heptafluorotantalate

Purity: 99.99% CAS No.: 16924-00-8 Molecular Formula: K2TaF7 Formula Weight: 392.14 Physical Form: Powder Density: 4.560 UN No.: 3288 MDL No.: MFCD00042369

Formula: F₇K₂Ta
Density: 4.560
Storage Temperature: Ambient
MDL Number: MFCD00042369
CAS Number: 16924-00-8
EINECS: 240-986-1
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR