Aluminium tri-sec-butanolate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide

Packaged under argon in resealable ChemSeal™ bottles.

43712-22 43712-36
CAAA43712-22EA 62.73 CAD
CAAA43712-22 CAAA43712-36
Aluminium tri-sec-butanolate ≥95%
Aluminium tri-sec-butanolate

Purity: 95% CAS No.: 2269-22-9 Molecular Formula: Al[OCH(CH3)CH2CH3]3 Formula Weight: 246.32 Flash Point: 27°(82°F) Physical Form: Liquid Density: 0.967 Refractive Index: 1.4390 Sensitivity: Moisture Sensitive Odor: Alcohol-like Boiling Point: 200-206°/30mm Beilstein Reference: 3910908 UN No.: 1992 MDL No.: MFCD00009327

Formula: C₁₂H₂₇AlO₃
MW: 246.33 g/mol
Boiling Pt: 200…206 °C (30 mmHg)
Density: 0.967
Flash Pt: 27 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009327
CAS Number: 2269-22-9
EINECS: 218-871-2
UN: 1992
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR