Hafnium diboride ≥99.5% (metals basis excluding Zr) (max. 2% Zr)

Supplier: Thermo Scientific Chemicals

Synonyms: Hafnium boride

43664-09 43664-18 43664-30
CAAA43664-09EA 121.03 CAD
CAAA43664-09 CAAA43664-18 CAAA43664-30
Hafnium diboride ≥99.5% (metals basis excluding Zr) (max. 2% Zr)
Hafnium diboride

Purity: 99.5% CAS No.: 12007-23-7 Molecular Formula: B2Hf Formula Weight: 200.11 Physical Form: -325 Mesh Powder Density: 10.5 Melting Point: 3250° MDL No.: MFCD00016125

Formula: B₂Hf
MW: 200.11
Melting Pt: 3250 °C
Density: 10.5
Storage Temperature: Ambient
MDL Number: MFCD00016125
CAS Number: 12007-23-7
EINECS: 234-500-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR