Disodium terephthalate 99+%

Supplier: Thermo Scientific Chemicals

42946-06 42946-14 42946-22 42946-36
CAAA42946-06EA 49.77 CAD
CAAA42946-06 CAAA42946-14 CAAA42946-22 CAAA42946-36
Disodium terephthalate 99+%
Disodium terephthalate

Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.


Purity: 99+% CAS No.: 10028-70-3 Molecular Formula: C6H4(COONa)2 Formula Weight: 210.10 Physical Form: Powder

Formula: C₈H₄Na₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00013137
CAS Number: 10028-70-3
EINECS: 233-075-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR