Zirconium(IV) oxide cement
Supplier: Thermo Scientific Chemicals
|
Warning
|
42493-30
42493-A1
CAAA42493-30EA
135.25
CAD
CAAA42493-30
CAAA42493-A1
CAAA42493-A7
Zirconium(IV) oxide cement
Zirconium (IV) oxide
One component adhesive. Bonds to Zirconium oxide and Silicon. Exhibit good dielectric properties
CAS No.: 1314-23-4
Physical Form: Liquid/paste
Appearance: Pale brown
Odor: Odorless
Merck Reference: 14,10180
MDL No.: MFCD01868884
Formula:
ZrO₂ Boiling Pt: 4300 °C (1013 hPa) Melting Pt: 2680 °C Density: 5.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS Number: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...