Lithium cobaltite (lithium cobalt(III) oxide) ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

42090-14 42090-22
CAAA42090-14EA 172.22 CAD
CAAA42090-14 CAAA42090-22
Lithium cobaltite (lithium cobalt(III) oxide) ≥99.5% (metals basis)
Lithium cobaltite (Lithium cobalt (III) oxide)

Purity: 99.5% CAS No.: 12190-79-3 Molecular Formula: LiCoO2 Formula Weight: 97.88 Physical Form: 5 Micron APS Powder Sensitivity: Hygroscopic MDL No.: MFCD00049786

Formula: CoLiO₂
Storage Temperature: Ambient
MDL Number: MFCD00049786
CAS Number: 12190-79-3
EINECS: 235-362-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR