Alizarin red S (sodium salt)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Alizarin sulfonic acid disodium salt, Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium
Transition interval: pH 4.6 (yellow) to pH 6.0 (red)
CAS No.: 130-22-3
Molecular Formula: C14H7O7SNa
Formula Weight: 342.27
Physical Form: Powder
Appearance: Yellow to red
Beilstein Reference: 4117090
Merck Reference: 14,8573
MDL No.: MFCD00013049
Formula:
C₁₄H₇NaO₇S Storage Temperature: Ambient |
MDL Number:
MFCD00013049 CAS Number: 130-22-3 EINECS: 204-981-8 Merck Index: 14,8573 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...