Iron(III) ethoxide ≥99.6% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41863-02
41863-04
41863-09
41863-18
CAAA41863-02EA
63.04
CAD
CAAA41863-02
CAAA41863-04
CAAA41863-09
CAAA41863-18
Iron(III) ethoxide ≥99.6% (metals basis)
Iron (III) ethoxide
Precursor for Sol-Gel processes
Soluble in anhydrous hexane, alcohols
Purity: 99.6%
CAS No.: 5058-42-4
Molecular Formula: C6H15FeO3
Formula Weight: 191.03
Physical Form: Powder/Chunk, Packaged under argon
Sensitivity: Moisture Sensitive
Appearance: Brown to dark grey
Odor: Odorless
Melting Point: 120°
Boiling Point: 155°/0.1mm
Solubility: Soluble in anhydrous hexane, alcohols
UN No.: 3181
MDL No.: MFCD00078028
Precursor for Sol-Gel processes
Formula:
C₆H₁₅FeO₃ Boiling Pt: 155 °C (0.1 mmHg) Melting Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078028 CAS Number: 5058-42-4 EINECS: 225-760-2 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...