Tungsten(VI) ethoxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
41795-03
41795-06
CAAA41795-03EA
119.45
CAD
CAAA41795-03
CAAA41795-06
Tungsten(VI) ethoxide
Tungsten (VI) ethoxide
Applications: Intermediate for sol-gel coatings
Soluble in ethyl acetate
CAS No.: 62571-53-3
Molecular Formula: C12H30O6W
Formula Weight: 454.21
Physical Form: Powder/chunks
Sensitivity: Moisture Sensitive
Appearance: Off-white to brown
Melting Point: 150°
Solubility: Soluble in ethyl acetate
UN No.: 3181
MDL No.: MFCD00156526
Intermediate for sol-gel coatings
Formula:
C₁₀H₂₅O₅W Melting Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156526 CAS Number: 62571-53-3 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...