Tungsten(V) ethoxide

Supplier: Thermo Scientific Chemicals
Warning

41794-04 41794-09
CAAA41794-04EA 170.64 CAD
CAAA41794-04 CAAA41794-09
Tungsten(V) ethoxide
Tungsten(V) ethoxide

Applications: Intermediate for sol-gel coatings


CAS No.: 26143-11-3 Molecular Formula: C10H25O5W Formula Weight: 409.15 Flash Point: 110°(230°F) Physical Form: Liquid Sensitivity: Moisture Sensitive Boiling Point: 110-115°/1mm MDL No.: MFCD00078041


Intermediate for sol-gel coatings

Formula: C₁₀H₂₅O₅W
Boiling Pt: 110…115 °C (1 mmHg)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00078041
CAS Number: 26143-11-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR