Aluminium triethanolate
Supplier: Thermo Scientific Chemicals
|
Danger
|
41336-09
41336-18
41336-30
CAAA41336-09EA
94.33
CAD
CAAA41336-09
CAAA41336-18
CAAA41336-30
Aluminium triethanolate
Aluminium triethanolate
CAS No.: 555-75-9
Molecular Formula: C6H15AlO3
Formula Weight: 162.16
Physical Form: Powder/ Chunks
Density: 1.142
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 154-159°
Boiling Point: 320°
Merck Reference: 14,338
UN No.: 2925
MDL No.: MFCD00009658
Formula:
C₆H₁₅AlO₃ Boiling Pt: 320 °C Melting Pt: 154…159 °C Density: 1.142 Storage Temperature: Ambient |
MDL Number:
MFCD00009658 CAS Number: 555-75-9 EINECS: 209-105-8 UN: 2925 ADR: 4.1,II Merck Index: 14,338 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...