Zinc chloride hydrate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

41247-09 41247-18 41247-30
CAAA41247-09EA 71.1 CAD
CAAA41247-09 CAAA41247-18 CAAA41247-30
Zinc chloride hydrate ≥99.99% (metals basis)
Zinc chloride hydrate

Purity: 99.99% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn.xH2O Formula Weight: 136.29(anhy) Physical Form: Crystalline Aggregates Appearance: White Odor: Odorless Merck Reference: 14,10132 UN No.: 3260 MDL No.: MFCD00011295

Formula: ZnCl₂
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 3260
ADR: 8,III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR