Fullerene C60 (C60/C70 70:28), powder, mixed, refined

Supplier: Thermo Scientific Chemicals
Warning

41181-03 41181-06 41181-MD
CAAA41181-03EA 323.9 CAD
CAAA41181-03 CAAA41181-06 CAAA41181-MD
Fullerene C60 (C60/C70 70:28), powder, mixed, refined
Fullerene C60

CAS No.: 99685-96-8 Appearance: Brown/black UN No.: 1325 MDL No.: MFCD00282904

Formula: C₆₀
Melting Pt: ? 280 °C
Density: ∼1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR