TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride
Reducing agent, specific for disulfides
Soluble in water
Purity: 98%
CAS No.: 51805-45-9
Molecular Formula: C9H15O6P.HCl
Formula Weight: 286.65
Physical Form: Crystalline powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 177-178°
Solubility: Soluble in water
Merck Reference: 14,9085
UN No.: 3261
MDL No.: MFCD00145469
Reducing agent, specific for disulfides
Formula:
C₉H₁₆ClO₆P MW: 286.65 g/mol Melting Pt: 177…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145469 CAS Number: 51805-45-9 UN: 3261 ADR: 8 II Merck Index: 14,9085 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...