Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilised, powder -100 Mesh

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

Stabilization: 4% Stabilised with calcia (CaO)

40452-18 40452-A1
CAAA40452-18EA 76.31 CAD
CAAA40452-18 CAAA40452-30 CAAA40452-A1
Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilised, powder -100 Mesh
Zirconium (IV) oxide

The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.


Plasma Spray Grade. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, Alcohols, Halogens.

Formula: O₂Zr
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2680
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR