Potassium heptafluorotantalate ≥99.7%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium heptafluorotantalate(V)

40316-18 40316-30
CAAA40316-18EA 301.78 CAD
CAAA40316-18 CAAA40316-30
Potassium heptafluorotantalate ≥99.7%
Potassium heptafluorotantalate

Decomposes in water. Slightly soluble in HF


Purity: 99.7% CAS No.: 16924-00-8 Molecular Formula: K2TaF7 Formula Weight: 392.14 Physical Form: Powder Density: 4.560 Solubility: Decomposes in water. Slightly soluble in HF UN No.: 3288 MDL No.: MFCD00042369

Formula: F₇K₂Ta
Density: 4.560
Storage Temperature: Ambient
MDL Number: MFCD00042369
CAS Number: 16924-00-8
EINECS: 240-986-1
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR