Potassium antimonate trihydrate ≥94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium pyroantimonate trihydrate, Potassium hexahydroxyantimonate trihydrate
39524-18
39524-30
CAAA39524-18EA
169.06
CAD
CAAA39524-18
CAAA39524-30
Potassium antimonate trihydrate ≥94%
Potassium antimonate trihydrate
Purity: 94+%
CAS No.: 12208-13-8
Molecular Formula: KSbO3.3H2O
Formula Weight: 262.90 (208.85anhy)
Physical Form: Powder or crystals
Appearance: White to cream
Odor: Odorless
UN No.: 1549
MDL No.: MFCD00150439
Formula:
H₆KO₆Sb MW: 262.90 g/mol (208.85 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150439 CAS Number: 12208-13-8 EINECS: 235-387-7 UN: 1549 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...