Magnesium lactate trihydrate ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Magnesium dilactate trihydrate, (±)-Lactic acid magnesium salt trihydrate, (±)-2-Hydroxypropionic acid magnesium salt trihydrate, DL-Lactic acid magnesium salt trihydrate, (±)-2-Hydroxypropanoic acid magnesium salt trihydrate

39369-14 39369-22
CAAA39369-14EA 87.22 CAD
CAAA39369-14 CAAA39369-22 CAAA39369-36
Magnesium lactate trihydrate ≥99%
Magnesium lactate trihydrate

Soluble in water, Slightly soluble in alcohol


Soluble in water, Slightly soluble in alcohol

Formula: (H₃CCH(OH)COO)₂Mg
MW: 256.49 g/mol (202.45 anhy)
Melting Pt: 41 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00156534
CAS Number: 18917-93-6
EINECS: 242-671-4
Merck Index: 14,5672

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR