Lithium salicylate, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lithium 2-hydroxybenzoate
39356-14
39356-22
39356-36
CAAA39356-14EA
55.62
CAD
CAAA39356-14
CAAA39356-22
CAAA39356-36
Lithium salicylate, tech.
Lithium salicylate
Soluble in water
CAS No.: 552-38-5
Molecular Formula: C7H5LiO3
Formula Weight: 144.05
Physical Form: Powder
Odor: Sweet
Melting Point: dec.
MDL No.: MFCD00045813
Formula:
C₇H₅LiO₃ Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00045813 CAS Number: 552-38-5 EINECS: 209-011-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...