Bis(dimethylphosphino)methane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39275-02
39275-04
CAAA39275-02EA
180.12
CAD
CAAA39275-02
CAAA39275-04
Bis(dimethylphosphino)methane 98%
Bis(dimethylphosphino)methane
Purity: 98%
CAS No.: 64065-08-3
Molecular Formula: C5H14P2
Formula Weight: 136.11
Physical Form: Liquid, Packaged under argon
Density: 0.86
Appearance: Colorless
Boiling Point: 42°/12mm
UN No.: 2845
MDL No.: MFCD00058731
Formula:
C₅H₁₄P₂ Boiling Pt: 42 °C (12 mmHg) Density: 0.86 Storage Temperature: Ambient |
MDL Number:
MFCD00058731 CAS Number: 64065-08-3 UN: 2845 ADR: 4.2,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...