1,2-Bis(diethylphosphino)ethane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39272-03
39272-06
CAAA39272-03EA
148.99
CAD
CAAA39272-03
CAAA39272-06
CAAA39272-MD
1,2-Bis(diethylphosphino)ethane 98%
1,2-Bis(diethylphosphino)ethane
Purity: 98%
CAS No.: 6411-21-8
Molecular Formula: C10H24P2
Formula Weight: 206.25
Physical Form: Liquid, Packaged under argon
Density: 0.884
Appearance: Colorless
Odor: Pungent
Boiling Point: 124-126°/10mm
UN No.: 2845
MDL No.: MFCD00015175
Formula:
C₁₀H₂₄P₂ MW: 206.245 g/mol Boiling Pt: 124…126 °C (10 mmHg) Density: 0.884 Storage Temperature: Ambient |
MDL Number:
MFCD00015175 CAS Number: 6411-21-8 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...