Tricalcium diarsenate ≥95%

Supplier: Thermo Scientific Chemicals
Danger

39219-18 39219-30
CAAA39219-18EA 165.9 CAD
CAAA39219-18 CAAA39219-30
Tricalcium diarsenate ≥95%
Tricalcium diarsenate

Purity: 95% CAS No.: 7778-44-1 Molecular Formula: Ca3(AsO4)2 Formula Weight: 398.03 Physical Form: -80 Mesh Powder Density: 3.62 Appearance: White Odor: Odorless UN No.: 1573 MDL No.: MFCD00049407

Formula: As₂Ca₃O₈
Density: 3.62
Storage Temperature: Ambient
MDL Number: MFCD00049407
CAS Number: 7778-44-1
EINECS: 231-904-5
UN: 1573
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR