Iodine monochloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
Soluble in water, alcohol, ether, carbon disulfide
Fieser: 1,502 8,261 10,212 11,268 18,193 19,176 20,200 21,233
CAS No.: 7790-99-0
Molecular Formula: ClI
Formula Weight: 162.36
Physical Form: Crystalline/Liquid
Density: 3.1822
Sensitivity: Moisture and Light Sensitive
Appearance: Dark red/black
Odor: Pungent
Melting Point: 25-27°
Boiling Point: 96-98°
Solubility: Soluble in water, alcohol, ether, carbon disulfide
Beilstein Reference: 3902972
Merck Reference: 14,5017
UN No.: 1792
MDL No.: MFCD00011354
Formula:
ClI Boiling Pt: 96…98 °C Melting Pt: 25…27 °C Density: 3.1822 Storage Temperature: Ambient |
MDL Number:
MFCD00011354 CAS Number: 7790-99-0 EINECS: 232-236-7 UN: 1792 ADR: 8,II Merck Index: 14,5017 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...