Starch, soluble modified (insolubles 0.01% max)

Supplier: Thermo Scientific Chemicals

36673-22 36673-36
CAAA36673-22EA 76.47 CAD
CAAA36673-22 CAAA36673-36
Starch, soluble modified (insolubles 0.01% max)
Starch, soluble

For iodometric titrations

Instantly soluble in water


CAS No.: 9005-84-9 Physical Form: Powder Odor: Odorless Solubility: Instantly soluble in water MDL No.: MFCD00082026

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR