Calcium-2-ethylhexanoate Ca 3-8% in 2-ethylhexanoic acid, Superconductor Grade

Supplier: Thermo Scientific Chemicals
Warning

36657-06 36657-14
CAAA36657-06EA 71.42 CAD
CAAA36657-06 CAAA36657-14 CAAA36657-22
Calcium-2-ethylhexanoate Ca 3-8% in 2-ethylhexanoic acid, Superconductor Grade
Calcium 2-ethylhexanoate

CAS No.: 136-51-6 Molecular Formula: C16H30CaO4 Formula Weight: 326.50 Physical Form: Liquid Odor: Weak, characteristic MDL No.: MFCD00014001

Formula: C₁₆H₃₀CaO₄
Storage Temperature: Ambient
MDL Number: MFCD00014001
CAS Number: 136-51-6
EINECS: 205-249-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR