Zirconium(IV) oxide ≥99% (metals basis excluding Hf) stabilized, powder -325 Mesh (max. 4% Hf)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

Stabilization: Stabilised with yttria (Y₂O₃)

36272-22 36272-36
CAAA36272-22EA 153.89 CAD
CAAA36272-22 CAAA36272-36
Zirconium(IV) oxide ≥99% (metals basis excluding Hf) stabilized, powder -325 Mesh (max. 4% Hf)
Zirconium (IV) oxide

Purity: 99% CAS No.: 1314-23-4 Molecular Formula: ZrO2 + 10-15% Y2O3 Physical Form: -325 Mesh Powder Appearance: White Odor: Odorless Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00145546

Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: ∼5000 °C
Melting Pt: ∼2700 °C
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR