Zinc chloride hydrate 99,999 Metals basis, Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

35782-09 35782-18 35782-30
CAAA35782-09EA 121.5 CAD
CAAA35782-09 CAAA35782-18 CAAA35782-30
Zinc chloride hydrate 99,999 Metals basis, Puratronic®
Zinc chloride hydrate

Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn.xH2O Formula Weight: 136.29(anhy) Physical Form: -10 Mesh Crystalline Aggregates Appearance: White Odor: Odorless Merck Reference: 14,10132 UN No.: 3260 MDL No.: MFCD00011295

Formula: Cl₂Zn·H₂O
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 3260
ADR: 8 III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR