Sodium hypochlorite (11 - 15% Cl₂) available chlorine in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

33369-36 33369-A3 33369-A9
CAAA33369-36EA 109.02 CAD
CAAA33369-36 CAAA33369-A3 CAAA33369-A9
Sodium hypochlorite (11 - 15% Cl₂) available chlorine in aqueous solution
Sodium hypochlorite

Typically 12 to 14% NaOCl


CAS No.: 7681-52-9 Molecular Formula: ClNaO Formula Weight: 74.44 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.2 Appearance: Clear yellow or green Merck Reference: 14,8628 UN No.: 1791 MDL No.: MFCD00011120

Formula: NaOCl
MW: 74.44 g/mol
Melting Pt: -6
Density: 1.2
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8 III
Merck Index: 14,8628

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR