Aluminium oxide cement ≥95% (SiO₂ ∼ 5%)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxideAlumina C

33201-A1 33201-A1
CAAA33201-A1EA 135.09 CAD
CAAA33201-A1 AA33201-A1
Aluminium oxide cement ≥95% (SiO₂ ∼ 5%)
Aluminium oxide

Contains 0.5% organic binder to increase dry strength

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Merck Index: 13,00355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR