Benzyldimethylhexadecylammonium chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cetalkonium chloride, N-Benzyl-N,N-dimethylhexadecan-1-aminium chloride
Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).
Purity: 95%
CAS No.: 122-18-9
Molecular Formula: C25H46ClN.xH2O
Formula Weight: 396.10(anhy)
Physical Form: Powder
Appearance: White
Melting Point: 54-58°
Beilstein Reference: 3802341
Merck Reference: 14,2017
UN No.: 3261
MDL No.: MFCD00011625
Formula:
C₂₅H₄₆ClN Melting Pt: 54…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011625 CAS Number: 122-18-9 EINECS: 204-526-3 UN: 3261 ADR: 8,III Merck Index: 14,2017 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...