Ammonium dihydrogenarsenate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
31493-18
31493-30
CAAA31493-18EA
470.84
CAD
CAAA31493-18
CAAA31493-30
Ammonium dihydrogenarsenate 98%
Ammonium dihydrogenarsenate
Purity: 98%
CAS No.: 13462-93-6
Molecular Formula: NH4H2AsO4
Formula Weight: 158.98
Physical Form: Crystalline
Density: 2.311
Refractive Index: 1.577
Melting Point: 300° dec.
UN No.: 1557
MDL No.: MFCD00064864
Formula:
H₆AsNO₄ Melting Pt: 300 °C (dec.) Density: 2.311 Storage Temperature: Ambient |
MDL Number:
MFCD00064864 CAS Number: 13462-93-6 EINECS: 236-667-1 UN: 1557 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...