Anthrone ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Anthracen-9(10H)-one

30739-14 30739-22
CAAA30739-14EA 133.68 CAD
CAAA30739-14 CAAA30739-22
Anthrone ACS
Anthrone

Used for determination of carbohydrates


CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Physical Form: Crystalline Sensitivity: Light Sensitive Melting Point: 155-158° Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187


Used for determination of carbohydrates

Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0
Merck Index: 14,691

Order Now

Specification Test Results

Melting point No more than a 5° range including 156°
Sensitivity to carbohydrates Pass Test
Absorbance of reagent solution Pass test.
Solubility in ethyl acetate Pass test.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR