Bis(dimethylamino)chlorophosphine
Supplier: Thermo Scientific Chemicals
|
Danger
|
30307-03
30307-06
CAAA30307-03EA
116.45
CAD
CAAA30307-03
CAAA30307-06
Bis(dimethylamino)chlorophosphine
Bis(dimethylamino)chlorophosphine
CAS No.: 3348-44-5
Molecular Formula: C4H12ClN2P
Formula Weight: 154.58
Physical Form: Liquid, Packaged under argon
Refractive Index: 1.5010
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless
Odor: Pungent
UN No.: 3265
MDL No.: MFCD00014862
Formula:
C₄H₁₂ClN₂P Storage Temperature: Refrigerator |
MDL Number:
MFCD00014862 CAS Number: 3348-44-5 EINECS: 222-100-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...