Chloro(diethyl)phosphine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethylchlorophosphine

30204-03 30204-06
CAAA30204-03EA 142.2 CAD
CAAA30204-03 CAAA30204-06
Chloro(diethyl)phosphine
Chloro(diethyl)phosphine

CAS No.: 686-69-1 Molecular Formula: C4H10ClP Formula Weight: 124.55 Flash Point: 18°(66°F) Physical Form: Liquid Density: 1.023 Refractive Index: 1.4750 Sensitivity: Air and Moisture Sensitive Appearance: Colorless to pale yellow Boiling Point: 131-132° UN No.: 2845 MDL No.: MFCD00013621

Formula: C₄H₁₀ClP
MW: 124.55 g/mol
Boiling Pt: 131…132 °C
Density: 1.023
Flash Pt: 18 °C (66 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013621
CAS Number: 686-69-1
EINECS: 211-689-4
UN: 2845
ADR: 4.2 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR