Nickel Aluminium ≥99.0% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

22890-09 22890-18 22890-30
CAAA22890-09EA 72.36 CAD
CAAA22890-09 CAAA22890-18 CAAA22890-30
Nickel Aluminium ≥99.0% (metals basis)
Nickel Aluminium

Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines


Purity: 99.0% min CAS No.: 12003-78-0 Molecular Formula: NiAl Physical Form: -100+325 Mesh Powder, Intermetallic Odor: Odorless

Formula: NiAl
Storage Temperature: Ambient
MDL Number: MFCD00167392
CAS Number: 12003-78-0
EINECS: 234-439-6
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR