Iron(II) methoxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
22552-04
22552-09
CAAA22552-04EA
485.06
CAD
CAAA22552-04
CAAA22552-09
Iron(II) methoxide 98%
Iron (II) methoxide
Purity: 98%
CAS No.: 7245-21-8
Molecular Formula: C2H6FeO2
Formula Weight: 117.92
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Green
Odor: Alcohol-like
UN No.: 3181
MDL No.: MFCD00061474
Formula:
C₂H₆FeO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00061474 CAS Number: 7245-21-8 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...