Tin(II) methoxide

Supplier: Thermo Scientific Chemicals
Danger

20897-06 20897-14
CAAA20897-06EA 257.54 CAD
CAAA20897-06 CAAA20897-14
Tin(II) methoxide
Tin (II) methoxide

CAS No.: 14794-99-1 Molecular Formula: Sn(OCH3)2 Formula Weight: 180.76 Physical Form: Powder Sensitivity: Moisture Sensitive Melting Point: 242-243° MDL No.: MFCD00015612

Formula: C₂H₆O₂Sn
Melting Pt: 242…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00015612
CAS Number: 14794-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR