Tin(II) ethoxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
20896-03
20896-06
CAAA20896-03EA
75.21
CAD
CAAA20896-03
CAAA20896-06
CAAA20896-14
Tin(II) ethoxide
Tin (II) ethoxide
Reacts with water
CAS No.: 14791-99-2
Molecular Formula: C4H10O2Sn
Formula Weight: 208.81
Physical Form: Solid/semi-solid
Sensitivity: Air and Moisture Sensitive
Melting Point: ca 200° dec.
MDL No.: MFCD00015653
Formula:
C₄H₁₀O₂Sn MW: 208.83 g/mol Melting Pt: ∼ 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00015653 CAS Number: 14791-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...