Lithium tetrachlorocuprate 0.1 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dilithium tetrachlorocuprate

20620-AE 20620-AP
CAAA20620-AEEA 138.09 CAD
CAAA20620-AE CAAA20620-AP
Lithium tetrachlorocuprate 0.1 M in THF
Lithium tetrachlorocuprate

CAS No.: 15489-27-7 Molecular Formula: Li2CuCl4 Formula Weight: 219.24 Physical Form: Liquid Density: 0.910 Sensitivity: Moisture Sensitive Appearance: Dark orange UN No.: 2056 MDL No.: MFCD00011081

Formula: Cl₄CuLi₂
MW: 219.24 g/mol
Density: 0.910
Storage Temperature: Ambient
MDL Number: MFCD00011081
CAS Number: 15489-27-7
UN: 2056
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR