Tantalum(V) methanolate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tantalum(V) methoxide
18839-06
18839-14
CAAA18839-06EA
279.66
CAD
CAAA18839-06
CAAA18839-14
Tantalum(V) methanolate
Tantalum(V) methanolate
CAS No.: 865-35-0
Molecular Formula: C5H15O5Ta
Formula Weight: 336.12
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 50°
Boiling Point: 189°/10mm
UN No.: 3181
MDL No.: MFCD00058817
Formula:
C₅H₁₅O₅Ta MW: 336.12 g/mol Boiling Pt: 189 °C (10 mmHg) Melting Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058817 CAS Number: 865-35-0 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...