Dibutyltin diacetate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di-n-butyltin diacetate
Common Applications: as Common Applications: as stabilizer in chlorinated organics, and Common Applications: as a catalyst for condensation reactions
Soluble in water and most organic solvents
Purity: 95%
CAS No.: 1067-33-0
Molecular Formula: C12H24O4Sn
Formula Weight: 351.01
Flash Point: 143°(290°F)
Physical Form: Liquid
Density: 1.32
Refractive Index: 1.4700
Appearance: Colorless to pale yellow
Odor: Acidic
Melting Point: 12°
Boiling Point: 142-145°/10mm
Solubility: Soluble in water and most organic solvents
UN No.: 2788
MDL No.: MFCD00008697
As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions
Formula:
C₁₂H₂₄O₄Sn MW: 351.03 g/mol Boiling Pt: 142…145 °C (10 mmHg) Melting Pt: 12 °C Density: 1.32 Flash Pt: 143 °C (290 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008697 CAS Number: 1067-33-0 EINECS: 213-928-8 UN: 2788 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...