Silver carbonate ∼ 0.7 mmole Ag₂CO₃/g on Celite

Supplier: Thermo Scientific Chemicals
Warning

18202-W5 18202-W9
CAAA18202-W5EA 352.34 CAD
CAAA18202-W5 CAAA18202-W9
Silver carbonate ∼ 0.7 mmole Ag₂CO₃/g on Celite
Silver carbonate

∼0.7 mmole Ag₂CO₃/g reagent


CAS No.: 534-16-7 Molecular Formula: Ag2CO3/Celite Physical Form: Powder Sensitivity: Light Sensitive Odor: Odorless MDL No.: MFCD00003403


Oxidizing agent for alcohols, diols/triols, hydroquinones, and amines.

Formula: CAg₂O₃
Melting Pt: 210…218 °C
Density: 5.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003403
CAS Number: 534-16-7
EINECS: 208-590-3
UN: 3077
ADR: 9,III
Merck Index: 12,08650

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR