Magnesium ethanolate Mg 21-22%

Supplier: Thermo Scientific Chemicals
Danger

14264-30 14264-A1
CAAA14264-30EA 191.18 CAD
CAAA14264-30 CAAA14264-A1
Magnesium ethanolate Mg 21-22%
Magnesium ethanolate

CAS No.: 2414-98-4 Molecular Formula: C4H10MgO2 Formula Weight: 114.44 Physical Form: Granular Density: 1.01 Sensitivity: Air and Moisture Sensitive Appearance: Off-white Odor: Odorless Melting Point: 270° dec. Beilstein Reference: 3678841 UN No.: 3205 MDL No.: MFCD00009060

Formula: C₄H₁₀MgO₂
Melting Pt: 270 °C (dec.)
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00009060
CAS Number: 2414-98-4
EINECS: 219-323-5
UN: 3205
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR