Lithium cobaltite (lithium cobalt(III) oxide) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14049-18
14049-30
14049-A1
CAAA14049-18EA
81.53
CAD
CAAA14049-18
CAAA14049-30
CAAA14049-A1
Lithium cobaltite (lithium cobalt(III) oxide) 97%
Lithium cobaltite (Lithium cobalt (III) oxide)
Purity: 97%
CAS No.: 12190-79-3
Molecular Formula: LiCoO2
Formula Weight: 97.88
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00049786
Formula:
CoLiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00049786 CAS Number: 12190-79-3 EINECS: 235-362-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...