Rubidium hydroxide hydrate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13481-06
13481-14
CAAA13481-06EA
208.56
CAD
CAAA13481-06
CAAA13481-14
Rubidium hydroxide hydrate ≥99% (metals basis)
Rubidium hydroxide hydrate
Applications: Oxidative chlorination catalyst
Soluble in water, alcohol
Purity: 99%
CAS No.: 12026-05-0
Molecular Formula: HORb.xH2O
Formula Weight: 102.48(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White to grey
Solubility: Soluble in water, alcohol
Merck Reference: 14,8285
UN No.: 2678
MDL No.: MFCD00011194
Oxidative chlorination catalyst
Formula:
RbOH Storage Temperature: Ambient |
MDL Number:
MFCD00011194 CAS Number: 1310-82-3 EINECS: 215-186-0 UN: 2678 ADR: 8,II Merck Index: 14,8285 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...