Chromium diboride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

12570-14 12570-22
CAAA12570-14EA 73.79 CAD
CAAA12570-14 CAAA12570-22
Chromium diboride ≥99% (metals basis)
Chromium diboride

Purity: 99% CAS No.: 12007-16-8 Molecular Formula: CrB2 Formula Weight: 73.62 Physical Form: -325 Mesh Powder Odor: Odorless MDL No.: MFCD00016043

Formula: B₂Cr
Storage Temperature: Ambient
MDL Number: MFCD00016043
CAS Number: 12007-16-8
EINECS: 234-499-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR