Potassium dicyanoaurate ≥99.96% (metals basis) Au ≥ 67.6%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gold(I) potassium cyanide, Potassium dicyanoaurate(I)
12552-03
12552-06
CAAA12552-03EA
406.06
CAD
CAAA12552-03
CAAA12552-06
Potassium dicyanoaurate ≥99.96% (metals basis) Au ≥ 67.6%, Premion®
Potassium dicyanoaurate
Purity: 99.96%
CAS No.: 13967-50-5
Molecular Formula: C2AuKN2
Formula Weight: 288.33
Physical Form: Crystalline powder
Density: 3.45
Appearance: White
Odor: Odorless
Melting Point: dec.
Beilstein Reference: 6235525
Merck Reference: 14,7628
UN No.: 1588
MDL No.: MFCD00011414
Formula:
C₂AuKN₂ MW: 288.1 g/mol Melting Pt: Dec. Density: 3.45 Storage Temperature: Ambient |
MDL Number:
MFCD00011414 CAS Number: 13967-50-5 EINECS: 237-748-4 UN: 1588 ADR: 6.1 II Merck Index: 14,7628 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...