Potassium dicyanoargentate (54.2% Ag) ≥99.9% (metals basis) Ag 54.2%

Supplier: Thermo Scientific Chemicals
Danger

12551-14 12551-22 12551-36
CAAA12551-14EA 161.16 CAD
CAAA12551-14 CAAA12551-22 CAAA12551-36
Potassium dicyanoargentate (54.2% Ag) ≥99.9% (metals basis) Ag 54.2%
Potassium dicyanoargentate

Applications: Silver plating
Soluble in water


Purity: 99.9% CAS No.: 506-61-6 Molecular Formula: C2AgKN2 Formula Weight: 199.01 Physical Form: Powder Density: 2.36 Appearance: White Solubility: Soluble in water Merck Reference: 14,7669 UN No.: 1588 MDL No.: MFCD00036297


Silver plating

Formula: C₂AgKN₂
Density: 2.36
Storage Temperature: Ambient
MDL Number: MFCD00036297
CAS Number: 506-61-6
EINECS: 208-047-0
UN: 1588
ADR: 6.1,I
Merck Index: 14,7669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR